Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TC-PTP/PTPN2 Antibody (001), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB316131 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316131 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316131 Supplier Novus Biologicals Supplier No. NBP290722JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TC-PTP/PTPN2 Monoclonal antibody specifically detects TC-PTP/PTPN2 in Mouse samples. It is validated for ELISA

Specifications

Antigen TC-PTP/PTPN2
Applications ELISA
Classification Monoclonal
Clone 001
Conjugate Janelia Fluor 549
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias protein tyrosine phosphatase, non-receptor type 2, PTPTPTN2, T-cell protein tyrosine phosphatase, T-cell protein-tyrosine phosphatase, TCELLPTP, TC-PTP, TCPTPEC 3.1.3.48, tyrosine-protein phosphatase non-receptor type 2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TC-PTP/PTPN2 (Uniprot#: Q06180-1; Ser2-Asn314).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5771
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.