Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC-S 7009
GREENER_CHOICE

Catalog No. p-7116078
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity and selective HIF-2α inhibitor

TC-S 7009 is a high affinity and selective HIF-2α inhibitor (Kd = 81 nM). Binds to the HIF-2α PAS-B domain; disrupts HIF-2α-ARNT heterodimerization. Exhibits >60-fold selectivity for HIF-2α over HIF-1α. Decreases HIF-2α DNA-binding and attenuates expression of HIF-2α target genes in vitro.

Specifications

Chemical Name or Material N-(3-Chloro-5-fluorophenyl)-4-nitro-2,1,3-benzoxadiazol-5-amine
CAS 1422955-31-4
Assay Percent Range >99%
Quantity 25 mg
Molecular Formula C12H6ClFN4O3
Formula Weight Observed MW: 308.65
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.