Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ TC-S 7010
Click to view available options
Quantity:
10 mg
50 mg
Description
TC-S 7010 is a potent and selective inhibitor of Aurora kinase A (IC50 = 3.4 nM); exhibits 1000-fold selectivity for Aurora kinase A over Aurora kinase B. Displays antiproliferative activity in HCT116 and HT29 cells (IC50 values are 0.19 and 2.9 μM respectively).
Specifications
Specifications
Chemical Name or Material | N-(2-Chlorophenyl)-4-[[2-[[4-[2-(4-ethyl-1-piperazinyl)-2-oxoethyl]phenyl]amino]-5-fluoro-4-pyrimidinyl]amino]-benzamide |
CAS | 1158838-45-9 |
Quantity | 10 mg |
Target | Aurora Kinase Inhibitors |
Molecular Formula | C31H31ClFN7O2 |
Formula Weight | 588.07 |
Purity | >99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction