Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TC10 Antibody (SR1384), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32204350UL
Description
TC10 Monoclonal antibody specifically detects TC10 in Human, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
TC10 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Western Blot, Immunofluorescence | |
SR1384 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
ARHQTC10A, ras homolog gene family, member Q, Ras-like protein family member 7A, Ras-like protein TC10, RAS-like, family 7, member A, rho-related GTP-binding protein RhoQ, TC10RASL7A | |
A synthesized peptide derived from human TC10 (Uniprot #: P17081) | |
50 μg | |
Signal Transduction | |
23433 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction