Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TC10 Antibody (SR1384), Novus Biologicals™
SDP

Catalog No. p-200005691 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258925 100 μg
NB260120 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258925 Supplier Novus Biologicals Supplier No. NBP322043100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TC10 Monoclonal antibody specifically detects TC10 in Human, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen TC10
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone SR1384
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias ARHQTC10A, ras homolog gene family, member Q, Ras-like protein family member 7A, Ras-like protein TC10, RAS-like, family 7, member A, rho-related GTP-binding protein RhoQ, TC10RASL7A
Host Species Rabbit
Immunogen A synthesized peptide derived from human TC10 (Uniprot #: P17081)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23433
Target Species Human, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.