Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TC21/R-Ras2 Antibody (EM-50), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB002643 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002643 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002643 Supplier Novus Biologicals Supplier No. NBP262216JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TC21/R-Ras2 Monoclonal specifically detects TC21/R-Ras2 in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).

Specifications

Antigen TC21/R-Ras2
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone EM-50
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Flow (Intracellular)
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ras-related protein R-Ras2, related RAS viral (r-ras) oncogene homolog 2, TC21Ras-like protein TC21, Teratocarcinoma oncogene
Gene Symbols RRAS2
Host Species Mouse
Immunogen Human TC21
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 22800
Test Specificity The antibody MEM-31 recognizes a conformationally-dependent epitope of CD8, a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. CD8 is a disulfide-linked dimer and exists as a CD8 alpha/alpha homodimer or CD8 alpha/beta heterodimer (each monomer approx. 32-34 kDa). The antibody does not react with formaldehyde-fixed cells; negative in Western Blotting application.
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.