Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCEAL1 Antibody (OTI2G10), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP274472V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274472V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274472V Supplier Novus Biologicals Supplier No. NBP274472V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCEAL1 Monoclonal specifically detects TCEAL1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TCEAL1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2G10
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Nuclear phosphoprotein p21/SIIR, p21, pp21, SIIRTCEA-like protein 1, transcription elongation factor A (SII)-like 1, transcription elongation factor A protein-like 1, Transcription elongation factor S-II protein-like 1
Gene Symbols TCEAL1
Host Species Mouse
Immunogen Full length recombinant protein of human TCEAL1 (NP_004771) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 9338.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.