Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCERG1L Antibody (05), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB328374 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB328374 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB328374 Supplier Novus Biologicals Supplier No. NBP305852APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCERG1L Monoclonal antibody specifically detects TCERG1L in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen TCERG1L
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 05
Conjugate APC
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 5730476P14Rik, FLJ38950, MGC126584, transcription elongation regulator 1-like, transcription elongation regulator 1-like protein
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with a synthetic peptide corresponding to the C-terminus of the Human TCERG1L.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256536
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.