Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCF4 Antibody (TCF4/1705), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB382548 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382548 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382548 Supplier Novus Biologicals Supplier No. NBP308236AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCF4 Monoclonal antibody specifically detects TCF4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen TCF4
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone TCF4/1705
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias BHLHB19, bHLHb19E2-2, Class B basic helix-loop-helix protein 19, Immunoglobulin transcription factor 2, ITF-2, ITF2PTHS, MGC149724, SEF-2, SEF2-1, SEF2-1A, SEF2-1B, SEF2MGC149723, SL3-3 enhancer factor 2, TCF-4, transcription factor 4
Host Species Mouse
Immunogen Recombinant human TCF4 protein fragment (around aa 365-671) (exact sequence is proprietary) (Uniprot: P15884 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6925
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.