Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCF7L1/TCF3 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB056476 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB056476 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB056476 Supplier Novus Biologicals Supplier No. NBP298907AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TCF7L1/TCF3 Polyclonal specifically detects TCF7L1/TCF3 in Human samples. It is validated for Western Blot.

Specifications

Antigen TCF7L1/TCF3
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias HMG box transcription factor 3, TCF3, TCF-3, transcription factor 7-like 1, transcription factor 7-like 1 (T-cell specific, HMG-box)
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human TCF7L1/TCF3.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 83439
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.