Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCF7L2 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NBP119083B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP119083B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP119083B Supplier Novus Biologicals Supplier No. NBP119083B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TCF7L2 Polyclonal specifically detects TCF7L2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TCF7L2
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9NQB0
Gene Alias HMG box transcription factor 4, hTCF-4, T-cell factor 4, T-cell factor-4 variant A, T-cell factor-4 variant B, T-cell factor-4 variant C, T-cell factor-4 variant D, T-cell factor-4 variant E, T-cell factor-4 variant H, T-cell factor-4 variant I, T-cell factor-4 variant J, T-cell factor-4 variant K, T-cell factor-4 variant L, T-cell factor-4 variant M, T-cell factor-4 variant X2, T-cell-specific transcription factor 4, TCF-4T-cell factor-4 variant F, TCF4T-cell factor-4 variant G, transcription factor 7-like 2, transcription factor 7-like 2 (T-cell specific, HMG-box)
Gene Symbols TCF7L2
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human TCF7L2 (within residues 150-200). [Swiss-Prot# Q9NQB0]
Molecular Weight of Antigen 55 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6934
Test Specificity Human. Immunogen sequence has 93% homology to mouse and bovine.
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.