Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF7 Antibody, Novus Biologicals™
SDP

Catalog No. NB6001399SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001399SS 0.025 mg
NB6001399 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB6001399SS Supplier Novus Biologicals Supplier No. NB6001399SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

TRAF7 Polyclonal specifically detects TRAF7 in Human, Mouse, Rat, Equine samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen TRAF7
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml. Use reported in scientific literature (Zotti et al (2011)), Immunoprecipitation reported in scientific literature (Zotti et al (2011))
Formulation PBS containing 0.05% BSA with 0.05% Sodium Azide
Gene Alias DKFZp586I021, E3 ubiquitin-protein ligase TRAF7, EC 6.3.2.-, MGC7807, RFWD1RING finger protein 119, RNF119RING finger and WD repeat-containing protein 1, TNF receptor-associated factor 7ring finger and WD repeat domain 1
Gene Symbols TRAF7
Host Species Rabbit
Immunogen Amino acids 334-356 of human TRAF7 protein were used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 84231
Test Specificity Human (isoforms CRA_a and CRA_c).
Target Species Human, Mouse, Rat, Equine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.