Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ TCH-165

Click to view available options
Quantity:
10 mg
50 mg
Description
TCH-165 is an enhancer of 20S proteasome assembly. TCH-165 regulates the dynamic equilibrium between the 20S and 26S proteasome complexes, favoring 20S-mediated protein degradation. TCH-165 enhances the proteolytic activity of the 20S proteasome by increasing substrate accessibility to the 20S catalytic chamber through 20S gate opening. TCH-165 enhances 20S-mediated degradation of IDPs, α-syn, and tau in vitro, and does not induce the degradation of structured proteins such as GAPDH. TCH-165 enhances c-MYC degradation by the 20S proteasome, which reduces c-MYC levels in vivo, and is effective in bortezomib (Cat. No. 7282) resistant cells. TCH-165 down-regulates c-MYC target genes and inhibits cancer cell proliferation and tumor growth in vivo.
Specifications
Specifications
| Chemical Name or Material | Ethyl (4 R,5 R)-rel-1-benzyl-5-(4-(benzylamino)phenyl)-2-(4-methoxyphenyl)-4-phenyl-4,5-dihydro-1 H-imidazole-4-carboxylate |
| CAS | 1446350-60-2 |
| Quantity | 10 mg |
| Target | Proteasome Modulators |
| Molecular Formula | C39H37N3O3 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction