Learn More
Description
Specifications
Specifications
| Antigen | TCL1A |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 43121 |
| Conjugate | PE |
| Dilution | Flow Cytometry |
| Gene Alias | Oncogene TCL1, Oncogene TCL-1, Protein p14 TCL1, T-cell leukemia/lymphoma 1A, T-cell lymphoma-1, T-cell lymphoma-1A, TCL1T-cell leukemia/lymphoma protein 1A |
| Gene Symbols | TCL1A |
| Host Species | Mouse |
| Immunogen | The immunogen for this antibody was TCL1. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
