Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCL1A Antibody (TCL1/2078), Biotin, Novus Biologicals™
SDP

Catalog No. NB008841 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB008841 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB008841 Supplier Novus Biologicals Supplier No. NBP275750B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCL1A Monoclonal specifically detects TCL1A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TCL1A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TCL1/2078
Conjugate Biotin
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias Oncogene TCL1, Oncogene TCL-1, Protein p14 TCL1, T-cell leukemia/lymphoma 1A, T-cell lymphoma-1, T-cell lymphoma-1A, TCL1T-cell leukemia/lymphoma protein 1A
Gene Symbols TCL1A
Host Species Mouse
Immunogen Recombinant human TCL1A protein fragment (around aa 2-109) (exact sequence is proprietary) (Uniprot: P56279)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity T-cell leukemia/Lymphoma Protein 1A (TCL1A), also known as p14TCL1 is a product of the TCL1 gene that is involved in T-cell prolymphocytic leukemia (TPLL). T-PLL is a rare form of mature T-cell leukemia, which is consistently associated with chromosomal rearrangements characterized by the juxtaposition of the TCRA locus on chromosome 14q11 and the TCL1A gene on 14q32.13. TCL1 is overexpressed in Burkitt lymphoma, the majority of AIDS-related non-Hodgkin lymphoma-designated immunoblastic plasmacytoid lymphoma, lymphoblastic lymphoma, chronic lymphocytic leukemia, mantle cell lymphoma, follicular lymphoma, diffuse large B-cell lymphoma, and primary cutaneous B-cell lymphoma.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.