Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCL1A Antibody (TCL1/2747R), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB380934 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380934 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380934 Supplier Novus Biologicals Supplier No. NBP308384UV
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TCL1A Monoclonal antibody specifically detects TCL1A in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen TCL1A
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TCL1/2747R
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias Oncogene TCL1, Oncogene TCL-1, Protein p14 TCL1, T-cell leukemia/lymphoma 1A, T-cell lymphoma-1, T-cell lymphoma-1A, TCL1T-cell leukemia/lymphoma protein 1A
Host Species Rabbit
Immunogen Recombinant human TCL1A protein fragment (around aa 2-109) (exact sequence is proprietary) (Uniprot: P56279)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8115
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.