Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCP1 alpha Antibody (23c), Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB12013501
Description
TCP1 alpha Monoclonal specifically detects TCP1 alpha in Human, Mouse, Rat, Canine, Chicken, Hamster, Plant, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| TCP1 alpha | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 2-5 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin | |
| TCP1 | |
| Recombinant mouse TCP-1 alpha (C-terminal fragment). | |
| 0.1 mg | |
| Cancer, Hypoxia, Membrane Trafficking and Chaperones | |
| 6950 | |
| Human, Mouse, Rat, Canine, Chicken, Hamster, Plant, Rabbit, Sheep | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 23c | |
| Unconjugated | |
| CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| This recognizes other proteins, most notably the p102B' COP subunit of Golgi coatomer. It does not react with human Hsp60 protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction