Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCP1 alpha Antibody (23c), Novus Biologicals™

Rat Monoclonal Antibody
$181.90 - $487.50
Specifications
| Antigen | TCP1 alpha |
|---|---|
| Clone | 23c |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2-5 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin |
| Applications | Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018069
![]() |
Novus Biologicals
NB120135010.025MG |
0.025 mg |
Each for $181.90
|
|
|||||
NB12013501
![]() |
Novus Biologicals
NB12013501 |
0.1 mg |
Each for $487.50
|
|
|||||
Description
TCP1 alpha Monoclonal specifically detects TCP1 alpha in Human, Mouse, Rat, Canine, Chicken, Hamster, Plant, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| TCP1 alpha | |
| 1.0 mg/mL | |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Cancer, Hypoxia, Membrane Trafficking and Chaperones | |
| CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha | |
| TCP1 | |
| IgG2c | |
| Protein G purified | |
| This recognizes other proteins, most notably the p102B' COP subunit of Golgi coatomer. It does not react with human Hsp60 protein. |
| 23c | |
| Western Blot 2-5 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Plant, Rabbit, Sheep, Hamster, Canine, Chicken, Human, Mouse, Rat | |
| 6950 | |
| Recombinant mouse TCP-1 alpha (C-terminal fragment). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title