Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCP10 Antibody (OTI2C2), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP274474C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274474C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274474C Supplier Novus Biologicals Supplier No. NBP274474C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCP10 Monoclonal specifically detects TCP10 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TCP10
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2C2
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias MGC34049, t-complex 10 (a murine tcp homolog), t-complex 10 (mouse), t-complex 10 homolog (mouse), T-complex protein 10A homolog, TCP10A
Gene Symbols TCP10
Host Species Mouse
Immunogen Full length recombinant protein of human TCP10 (NP_004601) produced in 293T cell.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6953.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.