Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCP11L2 Antibody (OTI2A10), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB274477APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274477APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274477APC Supplier Novus Biologicals Supplier No. NBP274477APC

Mouse Monoclonal Antibody

TCP11L2 Monoclonal specifically detects TCP11L2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TCP11L2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2A10
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias MGC40368, t-complex 11 (mouse) like 2, t-complex 11 (mouse)-like 2, T-complex protein 11-like protein 2
Gene Symbols TCP11L2
Host Species Mouse
Immunogen Full length recombinant protein of human TCP11L2 (NP_689985) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 255394.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.