Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR alpha Antibody (34), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP2271A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2271A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP2271A70 Supplier Novus Biologicals Supplier No. NBP252715AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCR alpha Monoclonal specifically detects TCR alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen TCR alpha
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 34
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA
Gene Symbols TRA
Host Species Mouse
Immunogen Purified recombinant soluble TCR from the T-cell hybridoma 85.33, corresponding to V alpha 11
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Cell Biology, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 6955.0
Test Specificity This antibody binds to the T-cell receptor alpha chains.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.