Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR alpha Antibody (34) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP25271525 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25271525 0.025 mg
NBP25271502 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25271525 Supplier Novus Biologicals Supplier No. NBP2527150.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCR alpha Monoclonal specifically detects TCR alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.

Specifications

Antigen TCR alpha
Applications Western Blot, Flow Cytometry, ELISA, CyTOF
Classification Monoclonal
Clone 34
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, CyTOF-ready
Formulation PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Gene Alias C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA
Host Species Mouse
Immunogen Purified recombinant soluble 85.33 scTCR.
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 6955
Test Specificity This antibody binds to the T-cell receptor alpha chains.
Target Species Human
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.