Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR alpha/beta Antibody (R73), FITC, Novus Biologicals™
SDP

Catalog No. NB355755 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355755 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355755 Supplier Novus Biologicals Supplier No. NBP212941F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCR alpha/beta Monoclonal antibody specifically detects TCR alpha/beta in Rat, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF

Specifications

Antigen TCR alpha/beta
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF
Classification Monoclonal
Clone R73
Conjugate FITC
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready
Formulation PBS
Gene Alias C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA
Host Species Mouse
Immunogen Rat T blasts and erythrocytes
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 6955
Target Species Rat, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.