Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR beta Antibody (H57-597), Alexa Fluor™ 647 - Chimeric, Novus Biologicals™
SDP

Catalog No. NB232573 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB232573 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB232573 Supplier Novus Biologicals Supplier No. NBP320118AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TCR beta Monoclonal antibody specifically detects TCR beta in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry

Specifications

Antigen TCR beta
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone H57-597
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias T cell receptor beta chain, TCRB, TCRbeta, Tib
Host Species Rabbit
Immunogen Hamsters were immunised with affinity purified DO-11.10 TCR, and produced detectable anti-TCR antibodies which were judged by the ability of the anti-sera to immunoprecipitate TCR from surface labelled DO-11.10 T cell hybridomas. Hamster 57 was selected for hybridisation, and B cell hybridomas were produced using polyethylene glycol fusion of hamster splenic B cells to the mouse myeloma variant. Selected mAb were screened after subsequent culturing for their ability to immunoprecipitate TCR from detergent lysates of surface-radiolabelled murine T cell hybridomas.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 21577
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.