Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR gamma/delta Antibody (11F2), PE, Novus Biologicals™
SDP

Catalog No. NB158543 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB158543 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB158543 Supplier Novus Biologicals Supplier No. NBP314621PE
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Human samples. It is validated for Flow Cytometry

Specifications

Antigen TCR gamma/delta
Applications Flow Cytometry
Classification Monoclonal
Clone 11F2
Conjugate PE
Dilution Flow Cytometry
Formulation PBS
Gene Alias T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG
Host Species Mouse
Immunogen Purified CD3 antigen complex
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 6965
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.