Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR gamma/delta Antibody (5A6.E9), Novus Biologicals™
SDP

Catalog No. NBP222489 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222489 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222489 Supplier Novus Biologicals Supplier No. NBP222489
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCR gamma/delta Monoclonal specifically detects TCR gamma/delta in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen TCR gamma/delta
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay
Classification Monoclonal
Clone 5A6.E9
Concentration 0.15 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1 - 2 ug, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional
Gene Accession No. P09693
Gene Alias T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG
Gene Symbols TRG
Host Species Mouse
Immunogen Human leukemic PEER cells
Purification Method Protein A purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 6965
Test Specificity This antibody reacts with Human.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.