Learn More
Description
Specifications
Specifications
| Antigen | TCR gamma/delta |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | B1.1 |
| Conjugate | DyLight 680 |
| Dilution | Flow Cytometry |
| Gene Alias | T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG |
| Gene Symbols | TRG |
| Host Species | Mouse |
| Immunogen | The immunogen for this antibody was gamma delta TCR. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
