Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR gamma/delta Antibody (GL-3), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB346596 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346596 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346596 Supplier Novus Biologicals Supplier No. FAB7297MFV450
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

TCR gamma/delta Monoclonal specifically detects TCR gamma/delta in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen TCR gamma/delta
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone GL-3
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, CyTOF-reported
Formulation 50mM Sodium Borate
Gene Alias T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG
Host Species Hamster
Immunogen Mouse intraepithelial lymphocytes
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 6965
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG2 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.