Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR V beta 5 Antibody (MEM-262), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB500520C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500520C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500520C Supplier Novus Biologicals Supplier No. NB500520C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCR V beta 5 Monoclonal specifically detects TCR V beta 5 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen TCR V beta 5
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone MEM-262
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias T cell receptor beta locus, T-cell antigen receptor, beta polypeptide, T-cell receptor, beta cluster, T-cell receptor, beta cluster, TCRB, TRB
Gene Symbols TRB
Host Species Mouse
Immunogen Human thymoma cell line HPB-ALL.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6957
Test Specificity The antibody MEM-262 recognizes beta chains of the TCR expressed by HPB-ALL cell line [carrying V(beta5.3)] and a small subset of peripheral blood T cells. This subset is larger than that recognized by other V(beta5.3)-specific antibodies.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.