Learn More
Description
Specifications
Specifications
| Antigen | TCR V beta 5 |
| Applications | Flow Cytometry, Immunoprecipitation, Functional Assay |
| Classification | Monoclonal |
| Clone | MEM-262 |
| Conjugate | PerCP |
| Dilution | Flow Cytometry |
| Gene Alias | T cell receptor beta locus, T-cell antigen receptor, beta polypeptide, T-cell receptor, beta cluster, T-cell receptor, beta cluster, TCRB, TRB |
| Gene Symbols | TRB |
| Host Species | Mouse |
| Immunogen | Human thymoma cell line HPB-ALL. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
