Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR V beta 5a Antibody (1C1), Novus Biologicals™
SDP

Catalog No. NBP222498 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222498 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222498 Supplier Novus Biologicals Supplier No. NBP222498
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCR V beta 5a Monoclonal specifically detects TCR V beta 5a in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen TCR V beta 5a
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Functional Assay
Classification Monoclonal
Clone 1C1
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional
Gene Accession No. P04435
Gene Alias T cell receptor beta locus, T-cell antigen receptor, beta polypeptide, T-cell receptor, beta cluster, T-cell receptor, beta cluster, TCRB, TRB
Gene Symbols TCB
Host Species Mouse
Immunogen Human TCR V beta 5(a)
Purification Method Protein G purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6957
Test Specificity This antibody reacts with Human and Primate.
Target Species Human, Primate
Content And Storage Store at 4C. Do not freeze.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.