Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR V delta 1 Antibody (TS8.2), Novus Biologicals™
SDP

Catalog No. NB404918 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
50 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB404918 50 ug
NBP222506 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB404918 Supplier Novus Biologicals Supplier No. NBP22250650UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen TCR V delta 1
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone TS8.2
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1 - 2 ug, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
Gene Alias T cell receptor delta locus, T-cell antigen receptor, delta polypeptide, T-cell receptor, delta (V,D,J,C), TCRD, TCRD, TRD@, TCRDV1, TRD@
Gene Symbols TRD
Host Species Mouse
Immunogen Human TCR V delta 1
Purification Method Protein G purified
Quantity 50 ug
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6964
Test Specificity This antibody reacts with Human.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.