Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR V delta 1 Antibody (TS8.2), Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $499.50
Specifications
Antigen | TCR V delta 1 |
---|---|
Clone | TS8.2 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 1 - 2 ug, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 |
Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
Description
TCR V delta 1 Monoclonal specifically detects TCR V delta 1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
TCR V delta 1 | |
0.2 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Human | |
6964 | |
Human TCR V delta 1 | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
TS8.2 | |
Flow Cytometry 1 - 2 ug, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
Monoclonal | |
Ascites | |
RUO | |
T cell receptor delta locus, T-cell antigen receptor, delta polypeptide, T-cell receptor, delta (V,D,J,C), TCRD, TCRD, TRD@, TCRDV1, TRD@ | |
TRD | |
IgG1 | |
Protein G purified | |
This antibody reacts with Human. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title