Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR V delta 2 Antibody (15D), Novus Biologicals™
SDP

Catalog No. NBP222508 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222508 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222508 Supplier Novus Biologicals Supplier No. NBP222508
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCR V delta 2 Monoclonal specifically detects TCR V delta 2 in Human, Drosophila, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen TCR V delta 2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay
Classification Monoclonal
Clone 15D
Concentration 0.15 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1 - 1:100, Flow Cytometry 1 - 2 ug, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional
Gene Alias T cell receptor delta locus, T-cell antigen receptor, delta polypeptide, T-cell receptor, delta (V,D,J,C), TCRD, TCRD, TRD@, TCRDV1, TRD@
Gene Symbols TRD
Host Species Mouse
Immunogen Human TCR V delta 2
Purification Method Protein A purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6964
Test Specificity This antibody reacts with Human. Drosophila reactivity reported in scientific literature (PMID: 18802083). Rhesus Monkey reactivity reported in scientific literature (PMID: 20660793).
Target Species Human, Drosophila, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.