Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR V delta 2 Antibody (B6), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB383252 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383252 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383252 Supplier Novus Biologicals Supplier No. NBP311706JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCR V delta 2 Monoclonal antibody specifically detects TCR V delta 2 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen TCR V delta 2
Applications Flow Cytometry
Classification Monoclonal
Clone B6
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias hDV102S1
Host Species Mouse
Immunogen Human recombinant TCR Vdelta2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28517
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.