Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TCS 1102
GREENER_CHOICESDP

Catalog No. 381810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-181-0 10 mg
38-185-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-181-0 Supplier Tocris Bioscience™ Supplier No. 3818/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, dual orexin receptor antagonist

Potent, dual orexin receptor antagonist (Ki values are 0.2 and 3 nM for OX2 and OX1 receptors respectively). Inhibits ADL-orexin B-mediated locomotion following i.p. administration in vivo and blocks orexin-A (Cat.No.1455) mediated increases in feeding behaviour. Brain penetrant.

Chemical Identifiers

CAS 916141-36-1
Molecular Formula C27H26N4O2S

Specifications

Quantity 10 mg
Formula Weight 470.59
Purity >98%
Molecular Weight (g/mol) 470.591
Chemical Name or Material TCS 1102
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.