Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TCS 359
GREENER_CHOICESDP

Catalog No. 259110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-911-0 10 mg
25-915-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-911-0 Supplier Tocris Bioscience™ Supplier No. 2591/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent FLT3 inhibitor

Potent inhibitor of FLT3 receptor tyrosine kinase (IC50 = 42 nM) that displays selectivity over a range of other kinases. Exhibits antiproliferative effects on MV4-11 cells, a human acute myelogenous leukemia cell line expressing a constitutively active mutant FLT3 (IC50 = 340 nM).

Chemical Identifiers

CAS 301305-73-7
Molecular Formula C18H20N2O4S

Specifications

Quantity 10 mg
Formula Weight 360.43
Purity >99%
Molecular Weight (g/mol) 360.428
Chemical Name or Material TCS 359
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.