Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDG Antibody, Novus Biologicals™
SDP

Catalog No. NBP220627 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220627 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220627 Supplier Novus Biologicals Supplier No. NBP220627
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TDG Polyclonal specifically detects TDG in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.

Specifications

Antigen TDG
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoassay
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:20000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Knockout Validated
Formulation PBS, 20% Glycerol (pH7) with 0.01% Thimerosal
Gene Accession No. Q13569
Gene Alias EC 3.2.2.29, thymine-DNA glycosylaseG/T mismatch-specific thymine DNA glycosylase
Gene Symbols TDG
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TDG. The exact sequence is proprietary.
Molecular Weight of Antigen 46 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Base Excision Repair, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 6996
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.