Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDO2 Antibody (998604), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB302322 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302322 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302322 Supplier Novus Biologicals Supplier No. FAB9768T

Mouse Monoclonal Antibody

TDO2 Monoclonal antibody specifically detects TDO2 in Human samples. It is validated for Immunohistochemistry, Flow Cytometry, CyTOF

Specifications

Antigen TDO2
Applications Immunohistochemistry, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 998604
Conjugate Alexa Fluor 594
Dilution Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias TDOTO, TPH2EC 1.13.11.11, TRPOtryptophanase, Tryptamin 2,3-dioxygenase, tryptophan 2,3-dioxygenase, Tryptophan oxygenase, Tryptophan pyrrolase, Tryptophanase
Host Species Mouse
Immunogen E. coli-derived human TDO2, Leu18-Phe388, Accession # P48775
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Lipid and Metabolism, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6999
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.