Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDP-43 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529949
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529949 Supplier Invitrogen™ Supplier No. PA529949
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, Neuro2A, GL261, NIH-3T3, BCL-1, Raw264.7, PC-12, Rat-2, Rat brain. Predicted reactivity: Mouse (95%), Rat (95%), Zebrafish (83%), Xenopus laevis (91%), Chicken (95%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TDP43 was first identified as a novel cellular protein that binds to HIV-1 virus TAR DNA sequence motifs and acts a transcriptional repressor to the HIV-1 LTR. Later experiments revealed that TDP43 also regulates the splicing of exon 9 of the cystic fibrosis transmembrane conductance regular (CFTR), most likely through the association with the UG repeats at the 3'e; splice site. A hyperphosphorylated, ubiquitinated, and cleaved form of TDP43 known as pathologic TDP43 is the major disease protein in ubiquitin-positive, tau-, and alpha-synuclein-negative frontotemporal dementia (FLTD-U). TDP43 is not related to TRBP1, and RNA binding protein that binds HIV-1 TAR RNA sequences.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDP-43
Applications ChIP sequencing (ChIP-seq), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, RNA Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene TARDBP
Gene Accession No. Q13148, Q921F2
Gene Alias 1190002A23Rik; ALS10; C85084; hypothetical protein; OTTHUMP00000002171; OTTHUMP00000002173; RCJMB04_5g9; RP4-635E18.2; TAR DNA binding protein; TAR DNA binding protein L homeolog; TAR DNA-binding protein 43; TAR DNA-binding protein-43; tardbp; tardbp.L; Tdp43; TDP-43; wu:fb77f02; wu:fc52g10; XELAEV_18035188mg
Gene Symbols TARDBP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TDP43. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230908, 23435, 298648
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.