Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDP-43/TARDBP Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11055376T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11055376T 0.025 mL
NB11055376 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11055376T Supplier Novus Biologicals Supplier No. NB11055376SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TDP-43/TARDBP Polyclonal specifically detects TDP-43/TARDBP in Human, Mouse, Chicken, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TDP-43/TARDBP
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, ELISA reported in scientific literature (PMID 25853864), Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:250
Formulation Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
Gene Accession No. Q13148
Gene Alias ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TDP43, TDP-43, TDP-43TAR DNA-binding protein 43
Gene Symbols TARDBP
Host Species Rabbit
Immunogen A synthetic peptide to a C-terminal region [within residues 350-414] of the human TARDBP protein. [Swiss-Prot: Q13148]
Molecular Weight of Antigen 45 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Epigenetics, Immunology, microRNA, Mitochondrial Fusion Proteins, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 23435
Target Species Human, Mouse, Chicken, Primate, Xenopus species, Zebrafish
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.