Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDP-43/TARDBP Antibody (DB9), Biotin, Novus Biologicals™
SDP

Catalog No. NBP250331B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250331B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250331B Supplier Novus Biologicals Supplier No. NBP250331B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TDP-43/TARDBP Monoclonal specifically detects TDP-43/TARDBP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen TDP-43/TARDBP
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone DB9
Conjugate Biotin
Dilution Western Blot, ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TDP43, TDP-43, TDP-43TAR DNA-binding protein 43
Gene Symbols TARDBP
Host Species Mouse
Immunogen A His-tagged recombinant protein corresponding to amino acid residues 208-414 of human TDP-43/TARDBP.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23435
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.