Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDP-43/TARDBP Antibody, Novus Biologicals™
SDP

Catalog No. NBP229648 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229648 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229648 Supplier Novus Biologicals Supplier No. NBP229648
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TDP-43/TARDBP Polyclonal specifically detects TDP-43/TARDBP in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TDP-43/TARDBP
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:250
Formulation PBS and 30% Glycerol with No Preservative
Gene Accession No. Q13148
Gene Alias ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TDP43, TDP-43, TDP-43TAR DNA-binding protein 43
Gene Symbols TARDBP
Host Species Rabbit
Immunogen Synthetic peptide C-terminal to the caspase-cleavage site (between D219 and V220) of human TDP-43. Accession # Q13148. Accession # Q13148
Molecular Weight of Antigen 43 kDa
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Epigenetics, Immunology, microRNA, Mitochondrial Fusion Proteins, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 23435
Test Specificity Specific for the ∼ 43kDa TDP-43 protein in Western blots of rat brain lysate. Expected molecular weight is dependent upon the TDP-43 species present in sample (full-length vs. truncated TDP-43). Under non-denaturing conditions (for example, by IHC), this antibody detects TDP-43 inclusions in human brain tissue with TDP-43 proteinopathy, but does not detect full-length nuclear TDP-43. Under denaturing conditions (for example, by Western Blot analysis) this antibody detects the C-terminus of full-length and truncated human TDP-43.
Target Species Human, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.