Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDRD1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP21148705 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP21148705 0.05 mg
NB018835 0.025 mg
NBP211487 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP21148705 Supplier Novus Biologicals Supplier No. NBP2114870.05MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TDRD1 Polyclonal specifically detects TDRD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen TDRD1
Applications Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.50 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:10000, ELISA 1:500, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:100
Formulation Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide
Gene Alias CT41.1Cancer/testis antigen 41.1, FLJ21082, tudor domain containing 1, tudor domain containing protein 1, tudor domain-containing protein 1
Gene Symbols TDRD1
Host Species Rabbit
Immunogen Synthetic peptides taken form an epitope lying between amino acids 578-601 with corresponding amino acid sequence. The synthetic peptide was convalently modified to achieve desired antigenicity before conjugating to a carrier protein.
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 56165
Test Specificity This antibody detects multiple bands of approximate MW of 83, 94, and a minor 114kDa in TDRD1 samples. The antibody does not cross reacts with other histones or DNA binding proteins
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.