Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TDRD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $670.50
Specifications
Antigen | TDRD1 |
---|---|
Dilution | Western Blot 1:10000, ELISA 1:500, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:100 |
Applications | Immunoprecipitation, Western Blot, ELISA, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018835
![]() |
Novus Biologicals
NBP2114870.025MG |
0.025 mg |
Each for $243.50
|
|
|||||
NBP21148705
![]() |
Novus Biologicals
NBP2114870.05MG |
0.05 mg |
Each for $487.50
|
|
|||||
NBP211487
![]() |
Novus Biologicals
NBP211487 |
0.1 mg |
Each for $670.50
|
|
|||||
Description
TDRD1 Polyclonal specifically detects TDRD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
TDRD1 | |
Immunoprecipitation, Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Human, Rat, Mouse | |
56165 | |
IgG | |
This antibody detects multiple bands of approximate MW of 83, 94, and a minor 114kDa in TDRD1 samples. The antibody does not cross reacts with other histones or DNA binding proteins |
Western Blot 1:10000, ELISA 1:500, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:100 | |
Polyclonal | |
Rabbit | |
CT41.1Cancer/testis antigen 41.1, FLJ21082, tudor domain containing 1, tudor domain containing protein 1, tudor domain-containing protein 1 | |
TDRD1 | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title