Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDRD3 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB291504 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291504 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291504 Supplier Novus Biologicals Supplier No. NBP146820PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TDRD3 Polyclonal antibody specifically detects TDRD3 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen TDRD3
Applications Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias FLJ21007, tudor domain containing 3, tudor domain-containing protein 3
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 400 and 450 of human tudor domain containing 3 using the numbering given in entry NP_110421.1 (GeneID 81550).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 81550
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.