Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TDRD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | TDRD3 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8677
![]() ![]() |
Proteintech Group Inc
133591AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Tudor domain-containing protein 3 (TDRD3) bears one tudor domain and an ubiquitin associated domain (UBA). Tudor domains interact with arginine-glycine-rich motifs in a methylarginine-dependent manner. TDRD3 has been shown to localize to cytoplasmic stress granules and may play a role in the regulation of mRNA translation.Specifications
TDRD3 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q66HC1, Q91W18, Q9H7E2 | |
219249, 306066, 81550 | |
TDRD3 Fusion Protein Ag4174 | |
Primary | |
-20°C | |
TDRD3 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
TDRD3, tudor domain containing 3 | |
TDRD3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title