Learn More
Description
Specifications
Specifications
| Antigen | TDRD9 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Alias | C14orf75, chromosome 14 open reading frame 75, DKFZp434N0820, EC 3.6.1, EC 3.6.4.13, FLJ36164, HIG-1, hypoxia-inducible HIG-1, MGC135025, NET54, putative ATP-dependent RNA helicase TDRD9, tudor domain containing 9, Tudor domain-containing protein 9 |
| Gene Symbols | TDRD9 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
