Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDRG1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP241197 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP241197 0.1 mg
NB404592 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP241197 Supplier Novus Biologicals Supplier No. NBP241197
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TDRG1 Polyclonal specifically detects TDRG1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen TDRG1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA
Formulation PBS with 0.02% Sodium Azide
Gene Alias LINC00532, Long Intergenic Non-Protein Coding RNA 532, Testis Development Related 1 (Non-Protein Coding)
Gene Symbols TDRG1
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid peptide near the center of human TDRG1. The immunogen is located within amino acids 40 - 90 of TDRG1. Amino Acid Squence: CIKISSTPPSQSRLDPQI
Molecular Weight of Antigen 11 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 732253
Test Specificity TDRG1 antibody is human reactive.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.