Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDRG1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB343784 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343784 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343784 Supplier Novus Biologicals Supplier No. NBP241197PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TDRG1 Polyclonal antibody specifically detects TDRG1 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen TDRG1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias LINC00532, Long Intergenic Non-Protein Coding RNA 532, Testis Development Related 1 (Non-Protein Coding)
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid peptide near the center of human TDRG1. The immunogen is located within amino acids 40 - 90 of TDRG1. Amino Acid Squence: CIKISSTPPSQSRLDPQI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 732253
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.